4-Fluor-3-iod-1H-indazol
Lieferant: ACTIVATE SCIENTIFIC
AS30604G1
AS30604G5
ACTIAS30604G1EA
0
CHF
ACTIAS30604G1
ACTIAS30604G5
4-Fluor-3-iod-1H-indazol
4-Fluoro-3-iodo-1H-indazole
Formel:
C₇H₄FIN₂ MW: 262.0200 g/mol |
MDL Number:
MFCD07781590 CAS-Nummer: 518990-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...