1H-Indazol-7-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS2546G1 AS2546G25 AS2546G5
ACTIAS2546G1EA 0 CHF
ACTIAS2546G1 ACTIAS2546G25 ACTIAS2546G5
1H-Indazol-7-carbonsäure
1H-Indazole-7-carboxylic acid
Formel: C₈H₆N₂O₂
MDL Number: MFCD06804572
CAS-Nummer: 677304-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR