1H-Indazol-6-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50660G1 AS50660G5
ACTIAS50660G1EA 0 CHF
ACTIAS50660G1 ACTIAS50660G5
1H-Indazol-6-carbonsäure
1H-Indazole-6-carboxylic acid
Formel: C₈H₆N₂O₂
MDL Number: MFCD06804571
CAS-Nummer: 704-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR