Methyl-1H-indazol-7-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS50740G1 AS50740G5
ACTIAS50740G1EA 0 CHF
ACTIAS50740G1 ACTIAS50740G5
Methyl-1H-indazol-7-carboxylat
Methyl-1H-indazol-7-carboxylat
Formel: C₉H₈N₂O₂
MDL Number: MFCD07371608
CAS-Nummer: 755752-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR