Ethyl-1H-indazol-5-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS1911G1 AS1911G5
ACTIAS1911G1EA 0 CHF
ACTIAS1911G1 ACTIAS1911G5
Ethyl-1H-indazol-5-carboxylat
Ethyl-1H-indazol-5-carboxylat
Formel: C₁₀H₁₀N₂O₂
MDL Number: MFCD03426220
CAS-Nummer: 192944-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR