Methyl-1H-indazol-4-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS50734G1 AS50734G5
ACTIAS50734G1EA 0 CHF
ACTIAS50734G1 ACTIAS50734G5
Methyl-1H-indazol-4-carboxylat
Methyl-1H-indazol-4-carboxylat
Formel: C₉H₈N₂O₂
MDL Number: MFCD07371610
CAS-Nummer: 192945-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR