3-Amino-7-chlor-5-nitro-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS22566G1 AS22566G25 AS22566G5
ACTIAS22566G1EA 0 CHF
ACTIAS22566G1 ACTIAS22566G25 ACTIAS22566G5
3-Amino-7-chlor-5-nitro-1H-indazol
3-Amino-7-chloro-5-nitro-1H-indazole
Formel: C₇H₅ClN₄O₂
MW: 212.5932 g/mol
MDL Number: MFCD07385614
CAS-Nummer: 1197193-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR