3-Amino-6-chlor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS22565G1 AS22565G25 AS22565G5
ACTIAS22565G1EA 0 CHF
ACTIAS22565G1 ACTIAS22565G25 ACTIAS22565G5
3-Amino-6-chlor-1H-indazol
3-Amino-6-chloro-1H-indazole
Formel: C₇H₆ClN₃
MW: 167.6000 g/mol
MDL Number: MFCD00047207
CAS-Nummer: 16889-21-7
EINECS: 240-925-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR