3-Amino-4-chlor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS33414G1 AS33414G5
ACTIAS33414G1EA 0 CHF
ACTIAS33414G1 ACTIAS33414G5
3-Amino-4-chlor-1H-indazol
3-Amino-4-chloro-1H-indazole
Formel: C₇H₆ClN₃
MDL Number: MFCD00663139
CAS-Nummer: 20925-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR