5-Chlor-3-iod-1H-indazol
Lieferant: ACTIVATE SCIENTIFIC
AS50939G1
AS50939G5
ACTIAS50939G1EA
0
CHF
ACTIAS50939G1
ACTIAS50939G5
5-Chlor-3-iod-1H-indazol
5-Chloro-3-iodo-1H-indazole
Formel:
C₇H₄ClIN₂ MW: 278.4800 g/mol |
MDL Number:
MFCD06739146 CAS-Nummer: 351456-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...