5-Amino-3-brom-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-1H-indazol-5-amine

AS2511G1 AS2511G5
ACTIAS2511G1EA 0 CHF
ACTIAS2511G1 ACTIAS2511G5
5-Amino-3-brom-1H-indazol
5-Amino-3-bromo-1H-indazole
Formel: C₇H₆BrN₃
MW: 212.500 g/mol
MDL Number: MFCD07781655
CAS-Nummer: 478837-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR