5-Brom-3-iod-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS8063G1 AS8063G5
ACTIAS8063G1EA 0 CHF
ACTIAS8063G1 ACTIAS8063G5
5-Brom-3-iod-1H-indazol
5-Bromo-3-iodo-1H-indazole
Formel: C₇H₄BrIN₂
MW: 322.9300 g/mol
MDL Number: MFCD07781637
CAS-Nummer: 459133-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR