6-Brom-4-fluor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS51010G1 AS51010G5
ACTIAS51010G1EA 0 CHF
ACTIAS51010G1 ACTIAS51010G5
6-Brom-4-fluor-1H-indazol
6-Bromo-4-fluoro-1H-indazole
Formel: C₇H₄BrFN₂
MW: 215,0200 g/mol
MDL Number: MFCD07781444
CAS-Nummer: 885520-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR