4-Brom-7-fluor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS14277G1 AS14277G5
ACTIAS14277G1EA 0 CHF
ACTIAS14277G1 ACTIAS14277G5
4-Brom-7-fluor-1H-indazol
4-Bromo-7-fluoro-1H-indazole
Formel: C₇H₄BrFN₂
MW: 215,0225 g/mol
MDL Number: MFCD16658655
CAS-Nummer: 1186334-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR