3-Brom-7-fluor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS31998G1 AS31998G5
ACTIAS31998G1EA 0 CHF
ACTIAS31998G1 ACTIAS31998G5
3-Brom-7-fluor-1H-indazol
3-Bromo-7-fluoro-1H-indazole
Formel: C₇H₄BrFN₂
MW: 215,0200 g/mol
MDL Number: MFCD13193285
CAS-Nummer: 1257853-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR