Methyl-6-brom-1H-indazol-3-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 6-bromo-1H-indazole-3-carboxylate

AS12266G1 AS12266G5
ACTIAS12266G1EA 0 CHF
ACTIAS12266G1 ACTIAS12266G5
Methyl-6-brom-1H-indazol-3-carboxylat
Methyl-6-brom-1H-indazol-3-carboxylat
Formel: C₉H₇BrN₂O₂
MW: 255,0700 g/mol
MDL Number: MFCD07371573
CAS-Nummer: 885278-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR