Methyl-5-brom-1H-indazol-3-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 5-bromo-1H-indazole-3-carboxylate

AS12263G1 AS12263G5
ACTIAS12263G1EA 0 CHF
ACTIAS12263G1 ACTIAS12263G5
Methyl-5-brom-1H-indazol-3-carboxylat
Methyl-5-brom-1H-indazol-3-carboxylat
Formel: C₉H₇BrN₂O₂
MDL Number: MFCD07371572
CAS-Nummer: 78155-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR