Methyl-3-brom-1H-indazol-4-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS1645G1 AS1645G5
ACTIAS1645G1EA 0 CHF
ACTIAS1645G1 ACTIAS1645G5
Methyl-3-brom-1H-indazol-4-carboxylat
Methyl-3-brom-1H-indazol-4-carboxylat
Formel: C₉H₇BrN₂O₂
MDL Number: MFCD03840642
CAS-Nummer: 885271-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR