7-Brom-3-chlor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS10178G1 AS10178G5
ACTIAS10178G1EA 0 CHF
ACTIAS10178G1 ACTIAS10178G5
7-Brom-3-chlor-1H-indazol
7-Bromo-3-chloro-1H-indazole
Formel: C₇H₄BrClN₂
MW: 231,4800 g/mol
MDL Number: MFCD06738285
CAS-Nummer: 885271-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR