6-Brom-3-chlor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS50940G1 AS50940G5
ACTIAS50940G1EA 0 CHF
ACTIAS50940G1 ACTIAS50940G5
6-Brom-3-chlor-1H-indazol
6-Bromo-3-chloro-1H-indazole
Formel: C₇H₄BrClN₂
MW: 231,48 g/mol
MDL Number: MFCD06738283
CAS-Nummer: 885271-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR