5-Brom-3-chlor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS1441G1 AS1441G5
ACTIAS1441G1EA 0 CHF
ACTIAS1441G1 ACTIAS1441G5
5-Brom-3-chlor-1H-indazol
5-Bromo-3-chloro-1H-indazole
Formel: C₇H₄BrClN₂
MW: 231,4800 g/mol
MDL Number: MFCD06738284
CAS-Nummer: 36760-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR