4-Brom-3-chlor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS10180G1 AS10180G5
ACTIAS10180G1EA 0 CHF
ACTIAS10180G1 ACTIAS10180G5
4-Brom-3-chlor-1H-indazol
4-Bromo-3-chloro-1H-indazole
Formel: C₇H₄BrClN₂
MW: 231,4800 g/mol
MDL Number: MFCD09026985
CAS-Nummer: 1000343-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR