1H-Indazol-6-boronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1H-Indazol-6-ylboronic acid, 6-Indazolyboronic acid, 1H-Indazol-6-yl-6-boronic acid

AS50898G1 AS50898G5
ACTIAS50898G1EA 0 CHF
ACTIAS50898G1 ACTIAS50898G5
1H-Indazol-6-boronsäure
1H-Indazole-6-boronic acid
Formel: C₇H₇BN₂O₂
MDL Number: MFCD04972419
CAS-Nummer: 885068-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR