1H-Indazol-5-boronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50899G1 AS50899G5
ACTIAS50899G1EA 0 CHF
ACTIAS50899G1 ACTIAS50899G5
1H-Indazol-5-boronsäure
1H-Indazole-5-boronic acid
Formel: C₇H₇BN₂O₂
MW: 161,9537 g/mol
MDL Number: MFCD06739151
CAS-Nummer: 338454-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR