1H-Indazol-4-boronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (1H-Indazol-4-yl)boronic acid, 1H-indazole-4-boronic acid

AS1452G1 AS1452G5
ACTIAS1452G1EA 0 CHF
ACTIAS1452G1 ACTIAS1452G5
1H-Indazol-4-boronsäure
1H-Indazole-4-boronic acid
Formel: C₇H₇BN₂O₂
MDL Number: MFCD09878901
CAS-Nummer: 1023595-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR