5-Brombenzoxazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromobenzo[d]oxazole

AS31507G1 AS31507G5
ACTIAS31507G1EA 0 CHF
ACTIAS31507G1 ACTIAS31507G5
5-Brombenzoxazol
5-Bromobenzoxazole
Formel: C₇H₄BrNO
MW: 198,0200 g/mol
MDL Number: MFCD03095026
CAS-Nummer: 132244-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR