3-Methoxypyridazin

Lieferant: ACTIVATE SCIENTIFIC

AS31788G1 AS31788G5
ACTIAS31788G1EA 0 CHF
ACTIAS31788G1 ACTIAS31788G5
3-Methoxypyridazin
3-Methoxypyridazine
Formel: C₅H₆N₂O
MW: 110.1139 g/mol
MDL Number: MFCD00234098
CAS-Nummer: 19064-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR