3-Pyridazincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50484G1 AS50484G5
ACTIAS50484G1EA 0 CHF
ACTIAS50484G1 ACTIAS50484G5
3-Pyridazincarbonsäure
Pyridazine-3-carboxylic acid
Formel: C₅H₄N₂O₂
MW: 124.10 g/mol
MDL Number: MFCD01646333
CAS-Nummer: 2164-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR