4,5-Dibrom-3(2H)-pyridazinon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4,5-Dibromopyridazin-3(2H)-one

AS31723G1 AS31723G5
ACTIAS31723G1EA 0 CHF
ACTIAS31723G1 ACTIAS31723G5
4,5-Dibrom-3(2H)-pyridazinon
4,5-Dibrom-3(2H)-pyridazinon
Formel: C₄H₂Br₂N₂O
MW: 253,89 g/mol
MDL Number: MFCD00023641
CAS-Nummer: 5788-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR