4-Brom-1-phenyl-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

AS22657G1 AS22657G5
ACTIAS22657G1EA 58 CHF
ACTIAS22657G1 ACTIAS22657G5
4-Brom-1-phenyl-1H-pyrazol
4-Bromo-1-phenyl-1H-pyrazole
Formel: C₉H₇BrN₂
MW: 223,0693 g/mol
MDL Number: MFCD00234505
CAS-Nummer: 15115-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR