5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS31612G1 AS31612G5
ACTIAS31612G1EA 0 CHF
ACTIAS31612G1 ACTIAS31612G5
5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbaldehyd
5-Chlor-1,3-dimethyl-1H-pyrazol-4-carbaldehyd
Formel: C₉H₁₁ClN₂O
MW: 158.59 g/mol
MDL Number: MFCD00052538
CAS-Nummer: 27006-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR