6-Chinoxalincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Quinoxaline-6-carboxylic acid

AS50624G1 AS50624G5
ACTIAS50624G1EA 0 CHF
ACTIAS50624G1 ACTIAS50624G5
6-Chinoxalincarbonsäure
6-Quinoxalinecarboxylic acid
Formel: C₉H₆N₂O₂
MW: 174,16 g/mol
MDL Number: MFCD01365834
CAS-Nummer: 6925-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR