5-Chinoxalincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Quinoxaline-5-carboxylic acid

AS1638G1 AS1638G5
ACTIAS1638G1EA 0 CHF
ACTIAS1638G1 ACTIAS1638G5
5-Chinoxalincarbonsäure
5-Quinoxalinecarboxylic acid
Formel: C₉H₆N₂O₂
MW: 174,1561 g/mol
MDL Number: MFCD02854494
CAS-Nummer: 6924-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR