(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride

AS33554G10 AS33554G100
ACTIAS33554G10EA 0 CHF
ACTIAS33554G10 ACTIAS33554G100
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure Hydrochlorid
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure Hydrochlorid
Formel: C₁₀H₁₂ClNO₂
MW: 213,6600 g/mol
MDL Number: MFCD00800357
CAS-Nummer: 77497-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR