Isochinolin-3-ylmethanol

Lieferant: ACTIVATE SCIENTIFIC

AS30906G1 AS30906G5
ACTIAS30906G1EA 0 CHF
ACTIAS30906G1 ACTIAS30906G5
Isochinolin-3-ylmethanol
Isochinolin-3-ylmethanol
Formel: C₁₀H₉NO
MDL Number: MFCD01859798
CAS-Nummer: 76884-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR