3-Chlor-6-(pyrrolidin-1-yl)pyridazin

Lieferant: ACTIVATE SCIENTIFIC

AS30171G1 AS30171G5
ACTIAS30171G1EA 0 CHF
ACTIAS30171G1 ACTIAS30171G5
3-Chlor-6-(pyrrolidin-1-yl)pyridazin
3-Chlor-6-(pyrrolidin-1-yl)pyridazin
Formel: C₈H₁₀ClN₃
MDL Number: MFCD05864735
CAS-Nummer: 66346-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR