Pyrazolo[1,5-a]pyridin

Lieferant: ACTIVATE SCIENTIFIC

AS8090G1 AS8090G5
ACTIAS8090G1EA 0 CHF
ACTIAS8090G1 ACTIAS8090G5
Pyrazolo[1,5-a]pyridin
Pyrazolo[1,5-a]pyridin
Formel: C₇H₆N₂
MW: 118,14 g/mol
MDL Number: MFCD08752622
CAS-Nummer: 274-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR