Pyrazolo[1,5-a]pyridin-5-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS14321G1 AS14321G5
ACTIAS14321G1EA 0 CHF
ACTIAS14321G1 ACTIAS14321G5
Pyrazolo[1,5-a]pyridin-5-carbonsäure
Pyrazolo[1,5-a]pyridin-5-carbonsäure
Formel: C₈H₆N₂O₂
MDL Number: MFCD12828626
CAS-Nummer: 104468-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR