Pyrazolo[1,5-a]pyridin-3-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS8092G1 AS8092G5
ACTIAS8092G1EA 0 CHF
ACTIAS8092G1 ACTIAS8092G5
Pyrazolo[1,5-a]pyridin-3-carbonsäure
Pyrazolo[1,5-a]pyridin-3-carbonsäure
Formel: C₈H₆N₂O₂
MDL Number: MFCD00229214
CAS-Nummer: 16205-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR