Methylpyrazolo[1,5-a]pyridin-2-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Pyrazolo[1,5-a]pyridine-2-carboxylic acid methyl ester

AS50579G1 AS50579G5
ACTIAS50579G1EA 0 CHF
ACTIAS50579G1 ACTIAS50579G5
Methylpyrazolo[1,5-a]pyridin-2-carboxylat
Methylpyrazolo[1,5-a]pyridin-2-carboxylat
Formel: C₉H₈N₂O₂
MDL Number: MFCD07367039
CAS-Nummer: 151831-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR