3-Brompyrazolo[1,5-a]pyridin

Lieferant: ACTIVATE SCIENTIFIC

AS36640G1 AS36640G5
ACTIAS36640G1EA 0 CHF
ACTIAS36640G1 ACTIAS36640G5
3-Brompyrazolo[1,5-a]pyridin
3-Brompyrazolo[1,5-a]pyridin
Formel: C₇H₅BrN₂
MW: 197.03 g/mol
MDL Number: MFCD06661764
CAS-Nummer: 5910-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR