3-Brom-1H-pyrazolo[3,4-b]pyridin

Lieferant: ACTIVATE SCIENTIFIC

AS12311G1 AS12311G25 AS12311G5
ACTIAS12311G1EA 0 CHF
ACTIAS12311G1 ACTIAS12311G25 ACTIAS12311G5
3-Brom-1H-pyrazolo[3,4-b]pyridin
3-Brom-1H-pyrazolo[3,4-b]pyridin
Formel: C₆H₄BrN₃
MW: 198,0200 g/mol
MDL Number: MFCD08669476
CAS-Nummer: 68618-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR