Methyl-5-methyl-4-oxazolcarboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS36233G1 AS36233G5
ACTIAS36233G1EA 0 CHF
ACTIAS36233G1 ACTIAS36233G5
Methyl-5-methyl-4-oxazolcarboxylat
Methyl-5-methyl-4-oxazolcarboxylat
Formel: C₆H₇NO₃
MDL Number: MFCD08689502
CAS-Nummer: 41172-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR