2,3-Dimethyl-6-nitro-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,3-dimethyl-6-nitro-2H-indazole

AS12292G25 AS12292G5
ACTIAS12292G25EA 0 CHF
ACTIAS12292G25 ACTIAS12292G5
2,3-Dimethyl-6-nitro-1H-indazol
2,3-Dimethyl-6-nitro-1H-indazole
Formel: C₉H₁₁N₃O₂
MW: 191,1867 g/mol
MDL Number: MFCD10703326
CAS-Nummer: 444731-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR