5-Brom-1,7-dimethyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-1,7-dimethylindazole

AS50941G1 AS50941G5
ACTIAS50941G1EA 0 CHF
ACTIAS50941G1 ACTIAS50941G5
5-Brom-1,7-dimethyl-1H-indazol
5-Bromo-1,7-dimethyl-1H-indazole
Formel: C₉H₉BrN₂
MW: 225,09 g/mol
MDL Number: MFCD11226595
CAS-Nummer: 1092352-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR