7-Methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS30839G1 AS30839G5
ACTIAS30839G1EA 0 CHF
ACTIAS30839G1 ACTIAS30839G5
7-Methyl-1H-indazol
7-Methyl-1H-indazole
Formel: C₈H₈N₂
MDL Number: MFCD05663434
CAS-Nummer: 3176-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR