6-Methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS8227G1 AS8227G5
ACTIAS8227G1EA 0 CHF
ACTIAS8227G1 ACTIAS8227G5
6-Methyl-1H-indazol
6-Methyl-1H-indazole
Formel: C₈H₈N₂
MW: 132,1625 g/mol
MDL Number: MFCD07781625
CAS-Nummer: 698-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR