5-Methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS1587G1 AS1587G5
ACTIAS1587G1EA 0 CHF
ACTIAS1587G1 ACTIAS1587G5
5-Methyl-1H-indazol
5-Methyl-1H-indazole
Formel: C₈H₈N₂
MW: 132,1600 g/mol
MDL Number: MFCD01606235
CAS-Nummer: 1776-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR