3-Methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS14092G1 AS14092G5
ACTIAS14092G1EA 0 CHF
ACTIAS14092G1 ACTIAS14092G5
3-Methyl-1H-indazol
3-Methyl-1H-indazole
Formel: C₈H₈N₂
MW: 132,1600 g/mol
MDL Number: MFCD00601361
CAS-Nummer: 3176-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR