3-Methyl-5-nitro-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS13354G1 AS13354G5
ACTIAS13354G1EA 0 CHF
ACTIAS13354G1 ACTIAS13354G5
3-Methyl-5-nitro-1H-indazol
3-Methyl-5-nitro-1H-indazole
Formel: C₈H₇N₃O₂
MW: 177,1601 g/mol
MDL Number: MFCD07082699
CAS-Nummer: 40621-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR